Learn More
Aviva Systems Biology Corp MCEE Antibody - C-terminal region (ARP60330_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60330_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCEE. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MCEE. Alternate names: GLOD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-009