Learn More
Active Motif MCF-7 nuclear extract (H2O2 post-treated)
SDP
Catalog No. NC1847772
Encompass
Active Motif's MCF-7 nuclear extract (H2O2 post-treated) was prepared from cell cultures of the human MCF-7 breast adenocarcinoma cancer cell line. The MCF-7 cell line was originally derived from a pleural effusion of a 69-year old Caucasian woman with metastatic breast cancer. MCF-7 cells are the best characterized and most commonly utilized cell lines in in vitro breast cancer studies. The MCF-7 cell line has retained several characteristics that are particular to mammary epithelium, including the ability to grow as a monolayer, form domes, and respond to hormones. These cells also display a low metastatic potential, leading to the assumption that they represent an early epithelial adenocarcinoma phenotype. These characteristics make MCF-7 cells an ideal […] For more information, please visit: https://www.activemotif.com/catalog/details/40810.
Catalog No. NC1847772 Supplier Active Motif Supplier No. 40810
Active Motif's MCF-7 nuclear extract (H2O2 post-treated) was prepared from cell cultures of the human MCF-7 breast adenocarcinoma cancer cell line. The MCF-7 cell line was originally derived from a pleural effusion of a 69-year old Caucasian woman with metastatic breast cancer. MCF-7 cells are the best characterized and most commonly utilized cell lines in in vitro breast cancer studies. The MCF-7 cell line has retained several characteristics that are particular to mammary epithelium, including the ability to grow as a monolayer, form domes, and respond to hormones. These cells also display a low metastatic potential, leading to the assumption that they represent an early epithelial adenocarcinoma phenotype. These characteristics make MCF-7 cells an ideal […] For more information, please visit: https://www.activemotif.com/catalog/details/40810.