Learn More
TRIBIOSCIENCE INC MCF10 Cell Complete Medium, 125 ML
SDP
Catalog No. NC3454602
Encompass_Preferred
Tribioscience’s MCF10 Cell Complete Medium is used for MCF10A cell growth and maintenance. This medium consists of all essential components and supplements for MCF10 cell culture. It is a ready-to-use medium for cell culture.
Catalog No. NC3454602 Supplier TRIBIOSCIENCE INC Supplier No. TBS8024125
Tribioscience’s MCF10 Cell Complete Medium is used for MCF10A cell growth and maintenance. This medium consists of all essential components and supplements for MCF10 cell culture. It is a ready-to-use medium for cell culture.