Learn More
ATCC MCF7; Breast Adenocarcinoma; Genomic DNA, Human
Catalog No. 502383419
Encompass
High molecular weight genomic DNA isolated from the MCF-7 human breast carcinoma cell line (ATCC HTB-22). The MCF7 line retains several characteristics of differentiated mammary epithelium including ability to process estradiol via cytoplasmic estrogen receptors and the capability of forming domes. Contains the Tx-4 oncogene.
Catalog No. 50-238-3419 Supplier ATCC Supplier No. HTB22D
High molecular weight genomic DNA isolated from the MCF-7 human breast carcinoma cell line (ATCC HTB-22). The MCF7 line retains several characteristics of differentiated mammary epithelium including ability to process estradiol via cytoplasmic estrogen receptors and the capability of forming domes. Contains the Tx-4 oncogene.