Learn More
ATCC MCF7; Breast Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502384386
Encompass
MCF7 are epithelial cells isolated from the breast tissue of a 69-year-old, White, female patient with metastatic adenocarcinoma. Use these cells in your breast cancer research.
Catalog No. 50-238-4386 Supplier ATCC Supplier No. HTB22
 Restricted Item
Add to Cart
Add to Cart
MCF7 are epithelial cells isolated from the breast tissue of a 69-year-old, White, female patient with metastatic adenocarcinoma. Use these cells in your breast cancer research.