Learn More
StatLab McFarland Standard Visual Comparison Set
SDP
Catalog No. 22130492
Change view
Click to view available options
5-level tube set
Catalog No.
22-130-492 5-level tube set
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-492 Supplier StatLab Supplier No. 2350
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
McFarland Standards Set
  • In preparing suspension for antimicrobial testing visually compare a suspension of bacteria in sterile saline or nutrient broth to the recommended McFarland Standard
  • Adjust the turbidity of bacterial suspensions so the number of bacteria will be within a specific range by referencing the McFarland Standards: 0.5,1, 2, 3, 4
  • Each tube contains 12 mL of suspension
  • 1 set
Specifications
Product Type McFarland Standard Set
Sterility Non-sterile
Quantity 1 Set