Learn More
CPC Scientific H-Asp-Phe-Asp-Met-Leu-Arg-Cys-Met-Leu-Gly-Arg-Val-Tyr-Arg-Pro-Cys-Trp-Gln-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MCH001A
Encompass
ONE-LETTER SEQUENCE: DFDMLRCMLGRVYRPCWQVMOLECULAR FORMULA: C105H160N30O26S4MOLECULAR WEIGHT:2386.88STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [128315-56-0], RESEARCH AREA: HormonalREFERENCES: J.M.Vaughan et al., Endocrinology, 125, 1660 (1989); F.Presse et al., Mol. Endocrinol., 4, 632 (1990); K.Takahashi et al., Ann. N. Y. Acad. Sci., 680, 619 (1993); J.-L.Nahon et al., Ann. N. Y. Acad. Sci., 680, 111 (1993)
Catalog No. 50-210-9719