Learn More
Eurofins Discoverx MCH(6-17), 500 μg. (92-1173)
Supplier:  Eurofins Discoverx 921173
Encompass
Melanin-concentrating Hormone (6-17) is a truncated version of a hypothalamic neuropeptide comprising amino acids 6 through 17. MCH is involved in feeding, mood, sleep-wake cycle, and energy balance. Overexpression is associated with obesity. Acts as an agonist of the melanin-concentrating hormone receptor (MCHR1). Provided as 500μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9701