Learn More
Bachem MCH (human, mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4025037.1000
Encompass_Preferred
MCH (human, mouse, rat) Trifluoroacetate, H-Asp-Phe-Asp-Met-Leu-Arg-Cys-Met-Leu-Gly-Arg-Val-Tyr-Arg-Pro-Cys-Trp-Gln-Val-OH, 128315-56-0, C105H160N30O26S4, Mr 2386.88, 1mg. Melanin-Concentrating Hormone (human, mouse, rat).
Catalog No. 50-259-505