Learn More
Cayman Chemical mCherry mRNACap-15mCTP 100ug
SDP
Supplier:  Cayman Chemical 39799100 UG
Encompass_Preferred
An mRNA for the expression of mCherry; encapsulation in LNPs can be used for mRNA delivery and expression of mCherry fluorescent protein in vitro or in vivo
Catalog No. 50-294-1986