Learn More
Cayman Chemical mCherry mRNACap-15mCTP 1mg
SDP
Catalog No. 502942225
Encompass_Preferred
An mRNA for the expression of mCherry; encapsulation in LNPs can be used for mRNA delivery and expression of mCherry fluorescent protein in vitro or in vivo
Catalog No. 50-294-2225 Supplier Cayman Chemical Supplier No. 397991 MG
An mRNA for the expression of mCherry; encapsulation in LNPs can be used for mRNA delivery and expression of mCherry fluorescent protein in vitro or in vivo