Learn More
Cayman Chemical mCherry mRNACap-1m1 100ug
SDP
Catalog No. 502943897
Encompass_Preferred
An mRNA for the expression of mCherry; encapsulation in LNPs can be used for mRNA delivery and expression of mCherry fluorescent protein in vitro or in vivo
Catalog No. 50-294-3897 Supplier Cayman Chemical Supplier No. 41962100 UG
An mRNA for the expression of mCherry; encapsulation in LNPs can be used for mRNA delivery and expression of mCherry fluorescent protein in vitro or in vivo