Learn More
Cayman Chemical mCherry mRNACap-1m1 1mg
SDP
Catalog No. 502943901
Encompass_Preferred
An mRNA for the expression of mCherry; encapsulation in LNPs can be used for mRNA delivery and expression of mCherry fluorescent protein in vitro or in vivo
Catalog No. 50-294-3901 Supplier Cayman Chemical Supplier No. 419621 MG
An mRNA for the expression of mCherry; encapsulation in LNPs can be used for mRNA delivery and expression of mCherry fluorescent protein in vitro or in vivo