Learn More
Eurofins Discoverx PathHunter MCHR1 Bioassay Kit, 10-Plate. (93-0940Y3-00150)
Catalog No. 502001623
Encompass
The PathHunter MCHR1 Bioassay kit provides a robust, highly sensitive, and easy-to-use functional cell-based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed for a complete assay, including cryopreserved cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure assay reproducibility and faster implementation from characterization to lot release. This bioassay has been optimized for a 96-well plate format. The protocol can be optimized further for running the assay in a high-throughput 384-well plate format. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-1623 Supplier Eurofins Discoverx Supplier No. 930940Y300150
Add to Cart
Add to Cart
Request a Quote
The PathHunter MCHR1 Bioassay kit provides a robust, highly sensitive, and easy-to-use functional cell-based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed for a complete assay, including cryopreserved cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure assay reproducibility and faster implementation from characterization to lot release. This bioassay has been optimized for a 96-well plate format. The protocol can be optimized further for running the assay in a high-throughput 384-well plate format. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.