Learn More
Eurofins Discoverx ChemiScreen MCH1 Melanin-concentrating Hormone Receptor Stable Cell Line, 2 vials. (HTS025C)
Catalog No. 502005692
Encompass
The ChemiScreen MCH1 Melanin-concentrating Hormone Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MCH) based activation of MCHR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses MCHR1 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of MCHR1, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5692 Supplier Eurofins Discoverx Supplier No. HTS025C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen MCH1 Melanin-concentrating Hormone Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MCH) based activation of MCHR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses MCHR1 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of MCHR1, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.