Learn More
MilliporeSigma MCHR1 MEMBRANE PREPARATION
Catalog No. 50818531
Encompass
Chemiscreen Membrane Preparation Human Recombinant MCHR1, 200 tests, For Radioligand binding assay, Reacts with Human, Maintain frozen at -70 deg.C for up to 2 years, Do not freeze and thaw
Catalog No. 50-818-531 Supplier MilliporeSigma Supplier No. HTS025M
Chemiscreen Membrane Preparation Human Recombinant MCHR1, 200 tests, For Radioligand binding assay, Reacts with Human, Maintain frozen at -70 deg.C for up to 2 years, Do not freeze and thaw