Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCI GEL™ CHP20P, MilliporeSigma™ Supelco™

Catalog No. 111009520 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Bottle
Bucket
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-100-9520 100 g Bottle
11-100-6672 500 g Bucket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9520 Supplier MilliporeSigma Supelco Supplier No. 13629U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MCI GEL™ CHP20P is a highly porous styrene-divinylbenzene polymer resin used as column packing material.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottle
Quantity 100 g
Product Line MCI GEL
Density 1.01 g/mL (at 25°C (true wet))

MCI GEL is a registered trademark of Mitsubishi Chemical Corp.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.