Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ McIllwain Tissue Chopper
SDP

Catalog No. 10000753 Shop All Stoelting Co Products
Change view
Click to view available options
Type:
Tissue Chopper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
10-000-753 Tissue Chopper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-000-753 Supplier Stoelting™ Supplier No. 51350
 Restricted Item
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Stoelting™ McIlwain Tissue Chopper is used for rapidly sectioning living or fixed tissue without freezing or embedding to harden the tissue.

  • Cuts living tissue into slices, cubes, arterial rings, etc. with a minimum of functional disturbance
  • Preparations can be used in studies of metabolism or for electrophysiology.
  • May also be used to cube fixed tissue for araldite embedding and EM.

Specifications

For Use With (Application) Studying Metabolism and Electrophysiology
Type Tissue Chopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.