Learn More
Boston Bioproducts Inc Citrate Phosphate Buffer (Mcilvaine Buffer, pH 6.0) - 500ml
Catalog No. NC1355356
Encompass
The Citrate-Phosphate buffer, also known as Mcllvaine Buffer, is composed of citric acid and sodium phosphate dibasic generating a pH range of 3.0-8.0. This buffer type can be used in various applications, including histology, medical diagnostics, immunohistochemistry, and molecular biology. By carefully adjusting the concentration of citrate ions and phosphate ions, optimal buffering capacity can be achieved, providing a controlled environment for sensitive biochemical processes.
Catalog No. NC1355356 Supplier Boston Bioproducts Inc Supplier No. BB2468500ML
The Citrate-Phosphate buffer, also known as Mcllvaine Buffer, is composed of citric acid and sodium phosphate dibasic generating a pH range of 3.0-8.0. This buffer type can be used in various applications, including histology, medical diagnostics, immunohistochemistry, and molecular biology. By carefully adjusting the concentration of citrate ions and phosphate ions, optimal buffering capacity can be achieved, providing a controlled environment for sensitive biochemical processes.