Learn More
McKesson General Medical MCKESSON OPA/28 HIGH-LEVEL DIS
Catalog No. NC1870725
Encompass_Preferred
McKesson OPA/28 High-Level Disinfectant MFG-73-OPA/28• Ready-to-use, no mixing or activating• Long, life, activated 28-day, 0.575% buffered ortho-Phthalaldehyde solutionwhich contains surfactants• 10 minute manual disinfection time at room temp of 68°F (20°C)• 5 minute AER disinfection time at 77°F (25°C)• Contraindicated for urological instrumentation to be utilized for cystoscopy orother urological procedures for patients with a history of bladder cancer• Contains buffering agents for corrosion inhibitors• Not made with natural rubber latex
This item is not returnable. View return policy
Catalog No. NC1870725 Supplier McKesson General Medical Supplier No. 852217
McKesson OPA/28 High-Level Disinfectant MFG-73-OPA/28• Ready-to-use, no mixing or activating• Long, life, activated 28-day, 0.575% buffered ortho-Phthalaldehyde solutionwhich contains surfactants• 10 minute manual disinfection time at room temp of 68°F (20°C)• 5 minute AER disinfection time at 77°F (25°C)• Contraindicated for urological instrumentation to be utilized for cystoscopy orother urological procedures for patients with a history of bladder cancer• Contains buffering agents for corrosion inhibitors• Not made with natural rubber latex