Learn More
Medchemexpress LLC HY-129700 5mg Medchemexpress, MCL-1/BCL-2-IN-2 CAS:2163793-44-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1297005MG
Encompass_Preferred
Medchemexpress, HY-129700 5mg MCL-1/BCL-2-IN-2 CAS:2163793-44-8 MCL-1/BCL-2-IN-2 (Compound 6) is a potent and selective Mcl-1 and Bcl-2 dual inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9815