Learn More
Medchemexpress LLC HY-129701 5mg Medchemexpress, MCL-1/BCL-2-IN-3 CAS:2163793-55-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1297015MG
Encompass_Preferred
Medchemexpress, HY-129701 5mg MCL-1/BCL-2-IN-3 CAS:2163793-55-1 MCL-1/BCL-2-IN-3 (Compound 2) is a potent and selective Mcl-1 and Bcl-2 dual inhibitor with IC50s of 5.95 and 4.78 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9816