Learn More
Cayman Chemical MCL0020trIfluoROCTAte sal 10mg
SDP
Catalog No. 502942949
Encompass_Preferred
An MC4R antagonist; selectively inhibits [Nle4,D-Phe7]-α-MSH binding to recombinant human MC4R over MC1R and MC3R (IC50s = 11.63, >10,000, and 1,115 nM, respectively); inhibits α-MSH-induced cAMP production in COS-1 cells expressing human MC4R from 0.1-100 µM; reduces stress-induced decreases in the time spent in the light area of the light/dark exploration test in mice; reverses restraint stress-induced decreases in food intake in rats
Catalog No. 50-294-2949 Supplier Cayman Chemical Supplier No. 4154410 MG
An MC4R antagonist; selectively inhibits [Nle4,D-Phe7]-α-MSH binding to recombinant human MC4R over MC1R and MC3R (IC50s = 11.63, >10,000, and 1,115 nM, respectively); inhibits α-MSH-induced cAMP production in COS-1 cells expressing human MC4R from 0.1-100 µM; reduces stress-induced decreases in the time spent in the light area of the light/dark exploration test in mice; reverses restraint stress-induced decreases in food intake in rats