Learn More
Medchemexpress LLC HY-16669 5mg Medchemexpress, Mcl1-IN-1 CAS:713492-66-1 Purity:>98%
SDP
Catalog No. 502081901
Encompass_Preferred
Medchemexpress, HY-16669 5mg Mcl1-IN-1 CAS:713492-66-1 Mcl1-IN-1 is an inhibitor of myeloid cell factor 1 (Mcl-1) (IC50=2.4 µM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1901 Supplier Medchemexpress LLC Supplier No. HY166695MG
Medchemexpress, HY-16669 5mg Mcl1-IN-1 CAS:713492-66-1 Mcl1-IN-1 is an inhibitor of myeloid cell factor 1 (Mcl-1) (IC50=2.4 µM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.