Learn More
Medchemexpress LLC HY-122627 5mg Medchemexpress, Mcl1-IN-8 CAS:678158-55-9 Purity:>98%
SDP
Catalog No. 501969634
Encompass_Preferred
Medchemexpress, HY-122627 5mg Mcl1-IN-8 CAS:678158-55-9 Mcl1-IN-8 (Comp8) is a Mcl-1-PUMA interface inhibitor, with a Ki of 0.3 μM. Mcl1-IN-8 (Comp8) exhibits dual activity on reduce PUMA-dependent apoptosis while deactivating Mcl-1-mediated anti-apoptosis in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9634 Supplier Medchemexpress LLC Supplier No. HY1226275MG
Medchemexpress, HY-122627 5mg Mcl1-IN-8 CAS:678158-55-9 Mcl1-IN-8 (Comp8) is a Mcl-1-PUMA interface inhibitor, with a Ki of 0.3 μM. Mcl1-IN-8 (Comp8) exhibits dual activity on reduce PUMA-dependent apoptosis while deactivating Mcl-1-mediated anti-apoptosis in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.