Learn More
Enzo Life Sciences Mcl-1 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP240F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Mcl-1. Source: Purified from rabbit serum. Species reactivity: Human Rabbit, Porcine, Monkey, Guinea pig, Dog. Applications: IF, IHC (PS), WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7158