Learn More
Cayman Chemical Mcl1L CytoplasmIc DomaInhu
SDP
Catalog No. 502949271
Encompass_Preferred
Source: Active recombinant human N-terminal His-tagged Mcl-1L expressed in E. coli • Amino acids: 2-327 • MW: 36 kDa
Catalog No. 50-294-9271 Supplier Cayman Chemical Supplier No. 33983100 UG
Add to Cart
Add to Cart
Source: Active recombinant human N-terminal His-tagged Mcl-1L expressed in E. coli • Amino acids: 2-327 • MW: 36 kDa