Learn More
Medchemexpress LLC HY-W013275 1mg Medchemexpress, MCLA hydrochloride CAS:128322-44-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0132751MG
Encompass_Preferred
Medchemexpress, HY-W013275 1mg MCLA hydrochloride CAS:128322-44-1 MCLA hydrochloride is a chemiluminescent reagent which can be used to quantify aqueous concentrations of superoxide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3347