Learn More
Cayman Chemical MCLAhydrochlorIde 10mg
SDP
Catalog No. 502966188
Encompass_Preferred
A chemiluminescent probe for superoxide production by activated leukocytes
Catalog No. 50-296-6188 Supplier Cayman Chemical Supplier No. 1867310 MG
A chemiluminescent probe for superoxide production by activated leukocytes