Learn More
Aviva Systems Biology Corp Mcm10 Antibody - N-terminal region (ARP36665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mcm10. It was validated on Western Blot. . Alternate names: 2410041F14Rik, AU018508, C330019M07Rik, C79164ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-509