Learn More
Aviva Systems Biology Corp MCM10 Antibody - middle region (ARP36656_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36656_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCM10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MCM10. Alternate names: CNA43, MGC126776, PRO2249, DNA43ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-56235