Learn More
Aviva Systems Biology Corp MCM2 Antibody - middle region (ARP36646_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36646_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MCM2. Alternate names: BM28, CCNL1, CDCL1, D3S3194, KIAA0030, MGC10606, MITOTIN, cdc19ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56227