Learn More
Aviva Systems Biology Corp MCM3 Antibody - C-terminal region (ARP36644_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36644_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MCM3. Alternate names: HCC5, MGC1157, P1-MCM3, P1.h, RLFBApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-53056