Learn More
Aviva Systems Biology Corp MCM4 Antibody - N-terminal region (ARP36662_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36662_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MCM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MCM4. Alternate names: CDC21, CDC54, hCdc21, P1-CDC21ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56238