Learn More
Aviva Systems Biology Corp MCM5 Antibody - N-terminal region (ARP36654_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36654_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MCM5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MCM5. Alternate names: CDC46, P1-CDC46ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56234