Learn More
Aviva Systems Biology Corp MCM6 Antibody - C-terminal region (ARP36650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCM6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MCM6. Alternate names: MCG40308, Mis5, P105MCMApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56230