Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4176MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: MCM7/1466. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: Raji or Ramos cells. Tonsil. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human MCM7 protein fragment (aa195-319). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 88kDa.Chromosome: 7q21.3-q22.1. Gene Symbol: MCM7. Gene ID: 4176. Ensemble: ENSG00000166508. Uniprot: NP_005907.
Catalog No. 50-188-4923