Learn More
Aviva Systems Biology Corp MCM7 Antibody - N-terminal region (ARP36653_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36653_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MCM7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MCM7. Alternate names: MCM2, CDC47, P85MCM, P1CDC47, PNAS146, P1.1-MCM3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56233