Learn More
Aviva Systems Biology Corp MCM8 Antibody - N-terminal region (ARP36694_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36694_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCM8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MCM8. Alternate names: C20orf154, MGC119522, MGC119523, MGC12866, MGC4816, REC, dJ967N21.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56254