Learn More
Electron Microscopy Sciences McMaster Egg Slide Chamber 1 each
SDP
Catalog No. 5027821
Encompass_Preferred
McMaster Egg Slide is used for determining fecal egg counts, an indicator of an animal's health.
Catalog No. 50-278-21 Supplier Electron Microscopy Sciences Supplier No. 6351275
McMaster Egg Slide is used for determining fecal egg counts, an indicator of an animal's health.