Learn More
Medchemexpress LLC HY-15578 10mg , McMMAF CAS:863971-19-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1557810MG
Encompass_Preferred
HY-15578 10mg McMMAF CAS: 863971-19-1 McMMAF is a protective group-conjugated MMAF. MMAF is a potent tubulin polymerization inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9533