Learn More
Aviva Systems Biology Corp MCOLN1 Antibody - N-terminal region (ARP35307_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35307_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCOLN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MCOLN1. Alternate names: ML4, MG-2, MLIV, MST080, TRPML1, MSTP080, TRP-ML1, TRPM-L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55693