Learn More
Medchemexpress LLC Mcoppb (trihydrochloride) | 1108147-88-1 | 100.0% | 50 MG
SDP
Catalog No. 5000339928
Encompass_Preferred
MCOPPB trihydrochloride is a nonpeptide nociceptin/orphanin FQ peptide (NOP)-receptor agonist, exhibiting a pKi of 10.07 for the human NOP receptor. It shows weaker activity at other opioid receptors. This compound is the trihydrochloride form of MCOPPB, making it suitable for various research applications.
  • Acts as a nociceptin receptor agonist
  • Demonstrates high purity of 99.98%
  • Stable under recommended storage conditions
  • Soluble in DMSO (100 mg/mL) for in vitro applications
  • Can be formulated for in vivo studies with clear solutions
Catalog No. 50-003-39928 Supplier Medchemexpress LLC Supplier No. HY1310150MG
MCOPPB trihydrochloride is a nonpeptide nociceptin/orphanin FQ peptide (NOP)-receptor agonist, exhibiting a pKi of 10.07 for the human NOP receptor. It shows weaker activity at other opioid receptors. This compound is the trihydrochloride form of MCOPPB, making it suitable for various research applications.
  • Acts as a nociceptin receptor agonist
  • Demonstrates high purity of 99.98%
  • Stable under recommended storage conditions
  • Soluble in DMSO (100 mg/mL) for in vitro applications
  • Can be formulated for in vivo studies with clear solutions