Learn More
Medchemexpress LLC MCOPPB trihydrochloride | 1108147-88-1 | 99.98% | 100 MG
SDP
Catalog No. 5000339892
Encompass_Preferred
MCOPPB trihydrochloride is a nociceptin receptor agonist with a pKi of 10.07, demonstrating weaker activity at other opioid receptors. It is the trihydrochloride form of MCOPPB, which is described as a new nonpeptide nociceptin/orphanin FQ peptide (NOP)-receptor agonist. It has a pKi of 10.07 ± 0.01 for the human NOP receptor. This product is for research use only.
  • Nociceptin receptor agonist
  • Weaker activity at other opioid receptors
  • Nonpeptide nociceptin/orphanin FQ peptide (NOP)-receptor agonist
  • For research use only
Catalog No. 50-003-39892 Supplier Medchemexpress LLC Supplier No. HY13101100MG
MCOPPB trihydrochloride is a nociceptin receptor agonist with a pKi of 10.07, demonstrating weaker activity at other opioid receptors. It is the trihydrochloride form of MCOPPB, which is described as a new nonpeptide nociceptin/orphanin FQ peptide (NOP)-receptor agonist. It has a pKi of 10.07 ± 0.01 for the human NOP receptor. This product is for research use only.
  • Nociceptin receptor agonist
  • Weaker activity at other opioid receptors
  • Nonpeptide nociceptin/orphanin FQ peptide (NOP)-receptor agonist
  • For research use only