Learn More
Medchemexpress LLC MCOPPB trihydrochloride | 1108147-88-1 | MFCD11840549 | 100.0% | 518.01 g·mol⁻1 | C26H43Cl3N4 | 5 MG
SDP
Catalog No. 5000263198
Encompass_Preferred
MCOPPB trihydrochloride is a research-grade nociceptin/orphanin FQ (NOP) receptor agonist supplied as a trihydrochloride salt. It exhibits high affinity for the human NOP receptor (pKi ≈ 10.07) and is intended for use in pharmacological studies, available in both solid and ready-to-use solution formats.
Catalog No. 50-002-63198 Supplier Medchemexpress LLC Supplier No. HY131015MG
MCOPPB trihydrochloride is a research-grade nociceptin/orphanin FQ (NOP) receptor agonist supplied as a trihydrochloride salt. It exhibits high affinity for the human NOP receptor (pKi ≈ 10.07) and is intended for use in pharmacological studies, available in both solid and ready-to-use solution formats.