Learn More
Cayman Chemical MCOPPBhydroCLOrID hydrate 10mg
SDP
Catalog No. 502954959
Encompass_Preferred
A potent, selective agonist for the nociceptin receptor (pKi = 10.1, EC50 = 0.39 nM); produces potent anxiolytic effects in rodents at 10 mg/kg, p.o. without inhibiting memory or motor function or inducing sedative side effects
Catalog No. 50-295-4959 Supplier Cayman Chemical Supplier No. 1203710 MG
A potent, selective agonist for the nociceptin receptor (pKi = 10.1, EC50 = 0.39 nM); produces potent anxiolytic effects in rodents at 10 mg/kg, p.o. without inhibiting memory or motor function or inducing sedative side effects