Learn More
Medchemexpress LLC MCP-1/CCL2 Human 2ug
SDP
Catalog No. 5000182943
Encompass_Preferred
MCP-1/CCL2 Protein Human is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein MCP-1/CCL2 Protein Human is a recombinant human MCP-1/CCL2(Q24-T99) expressed by E coil[1]
Catalog No. 50-001-82943 Supplier Medchemexpress LLC Supplier No. HYP72372UG
Add to Cart
Add to Cart
MCP-1/CCL2 Protein Human is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein MCP-1/CCL2 Protein Human is a recombinant human MCP-1/CCL2(Q24-T99) expressed by E coil[1]