Learn More
Medchemexpress LLC MCP-1/CCL2 Human H 50ug | 50ug
SDP
Catalog No. 5000194469
Encompass_Preferred
MCP-1/CCL2 Protein Human (HEK293 His-Avi) is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein MCP-1/CCL2 Protein Human (HEK293 His-Avi) is a recombinant human MCP-1/CCL2 (Q24-T99) expressed by HEK293 with His and Avi tags at the C-terminus[1]
Catalog No. 50-001-94469 Supplier Medchemexpress LLC Supplier No. HYP7848850UG
MCP-1/CCL2 Protein Human (HEK293 His-Avi) is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein MCP-1/CCL2 Protein Human (HEK293 His-Avi) is a recombinant human MCP-1/CCL2 (Q24-T99) expressed by HEK293 with His and Avi tags at the C-terminus[1]