Learn More
Medchemexpress LLC MCP-1/CCL2 Mouse 500ug | 500ug
SDP
Catalog No. 5000187314
Encompass_Preferred
MCP-1/CCL2 Protein Mouse is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein Mouse is a mouse MCP-1/CCL2 (Q24-R96) expressed by E coil[1]
Catalog No. 50-001-87314 Supplier Medchemexpress LLC Supplier No. HYP7764500UG
MCP-1/CCL2 Protein Mouse is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein Mouse is a mouse MCP-1/CCL2 (Q24-R96) expressed by E coil[1]