Learn More
Medchemexpress LLC MCP-1/CCL2 Mouse H 100ug
SDP
Catalog No. 5000182669
Encompass_Preferred
MCP-1/CCL2 Protein Mouse (HEK293) is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein Mouse (HEK293) is a mouse MCP-1/CCL2 (Q24-R96) expressed by HEK293[1]
Catalog No. 50-001-82669 Supplier Medchemexpress LLC Supplier No. HYP7235100UG
MCP-1/CCL2 Protein Mouse (HEK293) is a cytokine belonging to the CC chemokine family that interacts with the CCR2 chemokine receptor on the cell surface to mediate inflammatory immune responses viral infections and tumorigenesis MCP-1/CCL2 Protein Mouse (HEK293) is a mouse MCP-1/CCL2 (Q24-R96) expressed by HEK293[1]